missing translation for 'onlineSavingsMsg'
Learn More

IL-2 Rat anti-Human, PerCP-eFluor™ 710, Clone: MQ1-17H12, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-7029-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15589336

  • £102.00 / 25 tests

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

2 μm post- manufacturing filtered. Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications

Specifications

IL-2
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P60568
Interleukin-2, T-cell growth factor, TCGF, Aldesleukin
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MQ1-17H12
PerCP-eFluor™ 710
IL2
Liquid
IL2
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3558
Videos
Documents

For Research Use Only.