missing translation for 'onlineSavingsMsg'
Learn More

IL-2, PE, clone: MQ1-17H12, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7029-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15517086

  • £55.79 / 25 tests


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody.Applications Reported: MQ1-17H12 has been reported for use in intracellular flow cytometric analysis.Applications Tested: This MQ1-17H12 antibody is offered in 2 formats: -μg size: has been tested by intracellular staining of stimulated normal human perioheral blood cells. This can be used at less than or equal to 0.25μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated and tested by intracellular staining of stimulated normal human perioheral blood cells. This can be used at 5μL (0.125μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-2
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P60568
Interleukin-2, T-cell growth factor, TCGF, Aldesleukin
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MQ1-17H12
PE
IL2
Conjugated
IL2
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3558
Videos

For Research Use Only.