missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-2 R beta Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | IL-2 R beta |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL-2 R beta Polyclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Western Blot.Specifications
IL-2 R beta | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-15 R beta, IL-2 receptor subunit beta, IL2R beta, IL-2R subunit beta, IL2RB, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
IL2RB | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P16297 | |
3560 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-2 R beta Ala27-Glu240 Accession # P16297 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-2 R beta in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human IL-2 R beta is observed. |
For Research Use Only