missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Mouse anti-Cotton Rat, Clone: 144614, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | IL-2 |
---|---|
Clone | 144614 |
Applications | Western Blot |
Conjugate | Unconjugated |
Form | Purified |
Description
IL-2 Monoclonal specifically detects IL-2 in Cotton Rat samples. It is validated for Western Blot.Specifications
IL-2 | |
Western Blot | |
Purified | |
RUO | |
3558 | |
Monoclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
144614 | |
Unconjugated | |
Mouse | |
Cotton Rat | |
E. coli-derived recombinant cotton rat IL-2 Ala21-Gln155 Accession # AF398549 | |
Primary |
For Research Use Only