missing translation for 'onlineSavingsMsg'
Learn More

IL-19, clone: RN19, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-7191-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15208047

  • £156.97 / 500µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The RN19 antibody reacts with mouse interleukin-19 (IL-19), a member of the IL-10 family, which is secreted by activated keratinocytes and monocytes.Applications Reported: The RN19 antibody has been reported for use in ELISA capture of mouse IL-19.Applications Tested: The RN19 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse Interleukin-19 (IL-19) in combination with a biotinylated polyclonal anti-mouse IL-19 antibody for detection. A suitable range of concentrations of this antibody for ELISA capture is 1.0-4 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 4000 pg/mL - 30 pg/mL should be included in each ELISA plate.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-19
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q8CJ70
MDA-1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
ELISA
RN19
Unconjugated
Il19
Purified
Il19
IgG1, kappa
500μg
Store at 2-8°C.
Primary
329244
Videos

For Research Use Only.