missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18/IL-1F4 Propeptide Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | IL-18/IL-1F4 |
---|---|
Immunogen | E. coli-derived recombinant human IL-18/IL-1F4 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
IL-18/IL-1F4 Polyclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Western Blot, Simple Western, Knockout Validated.Specifications
IL-18/IL-1F4 | |
Goat | |
Polyclonal | |
IgG | |
Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot, Immunoassay | |
RUO |
E. coli-derived recombinant human IL-18/IL-1F4 | |
Primary | |
Human | |
Unconjugated | |
3606 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only