missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18 BPa Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | IL-18 BPa/IL18BP |
---|---|
Clone | 136031 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Description
IL-18 BPa/IL18BP Monoclonal specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
IL-18 BPa/IL18BP | |
LYOPH | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
O95998 | |
10068 | |
Mouse myeloma cell line NS0-derived recombinant human IL-18 BPa Thr31-Gly194 (predicted) Accession # O95998 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-18 BPa in ELISAs. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) IL-18, recombinant mouse (rm) IL-18 BPc and rmIL-18 BPd is observed. |
136031 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IL18 BPa, IL18BP, IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa | |
IL18BP | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only