Learn More
IL-17F Monoclonal Antibody (SHLR17), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£90.50 - £269.00
Specifications
Antigen | IL-17F |
---|---|
Clone | SHLR17 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15558636
|
Affymetrix eBioscience
25-7169-42 |
100 Tests |
£269.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15548636
|
Affymetrix eBioscience
25-7169-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The SHLR17 monoclonal antibody reacts with human interleukin (IL)-17F. IL-17F is a homodimeric, 34 kDa member of the IL-17 family, which includes IL-17(A), B, C, D, E (IL-25). Within this family, IL-17A and IL-17F are produced preferentially by T helper 17 (Th17) cells, with other members produced more widely. IL-17F and IL-17A have many overlapping functions, but knockout studies have shown each to have independent functions as well. \i{In vitro\i} cultured Th17 cells have been shown to express IL-17F and IL-17A homodimers and IL-17A/F heterodimers depending on the culture conditions and their differentiation state. A heterodimer of IL-17RA and IL-17RC has been shown to bind IL-17F, resulting in target cell-secretion of pro-inflammatory cytokines and chemokines, as well as neutrophil recruitment. Applications Reported: This SHLR17 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SHLR17 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of Th17 polarized human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation.
IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for Flow Cytometry.Specifications
IL-17F | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q96PD4 | |
IL17F | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL17F |
SHLR17 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Human | |
112744 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.