missing translation for 'onlineSavingsMsg'
Learn More

IL-17F, PE, clone: SHLR17, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7169-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15517106

  • £80.50 / 25 tests


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The SHLR17 monoclonal antibody reacts with human interleukin (IL)-17F. IL-17F is a homodimeric, 34kDa member of the IL-17 family, which includes IL-17(A), B, C, D, E (IL-25). Within this family, IL-17A and IL-17F are produced preferentially by T helper 17 (Th17) cells, with other members produced more widely. IL-17F and IL-17A have many overlapping functions, but knockout studies have shown each to have independent functions as well. In vitro cultured Th17 cells have been shown to express IL-17F and IL-17A homodimers and IL-17A/F heterodimers depending on the culture conditions and their differentiation state. A heterodimer of IL-17RA and IL-17RC has been shown to bind IL-17F, resulting in target cell-secretion of pro-inflammatory cytokines and chemokines, as well as neutrophil recruitment.Applications Reported: This SHLR17 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This SHLR17 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of TH17-polarized normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-17F
SHLR17
PE
Conjugated
Rat
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-17F
IgG1
RUO
Primary
Human
Videos