missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17E/IL-25 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | IL-17E/IL-25 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL-17E/IL-25 Polyclonal specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blot.Specifications
IL-17E/IL-25 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
IL25 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_542767 | |
64806 | |
E. coli-derived recombinant mouse IL-17E/IL-25 Val17-Ala169 Accession # NP_542767 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-17E/IL-25 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) IL-17E and recombinant rat IL-17E is observed, and less than 1% cross-reactivity with recombinant mouse (rm) IL-17, rmIL-17B, rhIL-17C, rhIL-17D, and rhIL-17F is observed. |
For Research Use Only