missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17A, Functional Grade, clone: eBio17CK15A5, eBioscience™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-7175-81
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The eBio17CK15A5 antibody reacts with mouse and rat IL-17A. Interleukin-17A (IL-17A) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. The cDNA encoding human IL-17A was isolated from a library of CD4+ T cells; the encoded protein exhibits 72 percent amino acid identity with HVS13 , an open reading frame from a T lymphotropic Herpesvirus saimiri, and 63 percent with mouse CTLA-8 (cytotoxic T-lymphocyte associated antigen-8). Human IL-17A exists as glycosylated 20-30 kD homodimers. High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells. IL-17A enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. Human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse IL-17A receptor. IL-23-dependent, IL-17A-producing CD4+ T cells (Th-17 cells) have been identified as a unique subset of Th cells that develops along a pathway that is distinct from the Th1- and Th2- cell differentiation pathways. The hallmark effector molecules of Th1 and Th2 cells, e.g., IFN-gamma and IL-4, have each been found to negatively regulate the generation of these Th-17 cells.Applications Reported: The eBio17CK15A5 antibody has been reported useful for mouse IL-17A ELISA and ELISPOT capture.Applications Tested: The Functional Grade Purified eBio17CK15A5 antibody has been tested by LAL assay to verify low endotoxin level and has been tested for mouse IL-17A ELISPOT and ELISA capture. The Functional Grade Purified eBio17CK15A5 antibody has been tested as the capture antibody in a sandwich ELISA for measurement of mouse IL-17A protein levels, in combination with the biotinylated eBio17B7 antibody (13-7177) for detection and recombinant mouse IL-17A (14-8171) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1.0 - 4.0 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate..Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.Specifications
IL-17A | |
Monoclonal | |
1mg/mL | |
PBS with no preservative; pH 7.2 | |
Q62386, Q61453 | |
Interleukin-17 | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Murine |
Dot blot | |
eBio17CK15A5 | |
Functional Grade | |
Il17a | |
Purified | |
Il17a | |
IgG2a, kappa | |
50μg | |
Store at 2-8°C. | |
Primary | |
16171 |
For Research Use Only.