missing translation for 'onlineSavingsMsg'
Learn More

IL-15, Functional Grade, clone: ct2nu, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-0157-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15268387

  • £176.24 / 500µg


Estimated Shipment
26-07-2018
Add to basket

Description

Description

Description: The ct2nu monoclonal antibody reacts with human IL-15 and neutralizes its bioactivity. IL-15, or IL-T, is a 14kDa proinflammatory cytokine and member of the IL-2 family. It plays a role in the development and maturation of macrophages and dendritic cells, and is essential to the proliferation and survival of NK cells and CD8^+ T cells. IL-15 is expressed by antigen-presenting cells, epithelial cells, and stromal cells. IL-15 activates the Jak/STAT pathway through its heterotrimeric receptor. Two subunits of this receptor, the beta and gamma chains, are shared with the rest of the family, while the alpha subunit is unique to IL-15. In mice, the alpha chain is required to chaperone IL-15 from the ER to secretion. This chaperoning is not necessary in humans, and free IL-15 is detectable in human biological samples. However, the receptor-cytokine complex has been indentified in humans and demonstrates agonistic activity several-fold higher than IL-15 alone.Applications Reported: The monoclonal antibody ct2nu reacts with and inhibits the bioactivity of human IL-15.Applications Tested: The ND50 of this antibody, as measured by inhibition of proliferation in CTLL-2 cells, is 1-10 ng/mL in the presence of 0.5 ng/mL recombinant human IL-15. Neutralization dose will vary based on cell type, cytokine concentration, and assay method. This antibody should be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

IL-15
Monoclonal
1mg/mL
PBS with no preservative; pH 7.2
P40933
Interleukin-15, MGC9721
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Cytokine Neutralization
ct2nu
Functional Grade
IL15
Purified
IL15
IgG1, kappa
500μg
Store at 2-8°C. Avoid repeated freeze/thaw cycles. KEEP CONTENTS STERILE.
Primary
3600
Videos

For Research Use Only.