missing translation for 'onlineSavingsMsg'
Learn More

IL-13, PE-Cyanine7, clone: eBio13A, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-7133-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15528636

  • £70.66 / 25µg


EU Stock 3
Add to basket

Description

Description

Description: The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12)Applications Reported: This eBio13A antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This eBio13A antibody has been tested by intracellular staining of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-13
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P20109
Interleukin-13, ALRH antibody, NC30 antibody, P600 antibody
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow CytometryþFlow Cytometry
eBio13A
PE-Cyanine7
Il13
Conjugated
Il13
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
16163
Videos

For Research Use Only.