missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13, Alexa Fluor 488, clone: eBio13A, eBioscience™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 53-7133-80
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12)Applications Reported: This eBio13A antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This eBio13A antibody has been tested by intracellular staining and flow cytometric analysis of Th2 polarized CD4 cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
IL-13 | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P20109 | |
Interleukin-13, ALRH antibody, NC30 antibody, P600 antibody | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Murine |
Flow Cytometry | |
eBio13A | |
Alexa Fluor 488 | |
Il13 | |
Conjugated | |
Il13 | |
IgG1, kappa | |
25μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
16163 |
For Research Use Only.