missing translation for 'onlineSavingsMsg'
Learn More

IL-13 Mouse anti-Human, PE, Clone: 85BRD, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7136-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15577096

  • £85.25 / 25 tests

Estimated Shipment
29-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Specifications

Specifications

IL-13
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P35225
Interleukin-13, ALRH antibody, NC30 antibody, P600 antibody
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
85BRD
PE
IL13
Liquid
IL13
IgG2b, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3596
Videos
Documents

For Research Use Only.