missing translation for 'onlineSavingsMsg'
Learn More

IL-13, PE, clone: 85BRD, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7136-42

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15587096

  • £190.00 / 100 tests

Estimated Shipment
30-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The 85BRD monoclonal antibody reacts with human interleukin-13 (IL-13). IL-13, also known as NC30, is a 12.5kDa protein secreted by activated T helper cells, CD8+ T cells, and NK cells. The biological activities of IL-13 include suppression of macrophage cytotoxic activity, upregulation of IL-1RA expression and suppression of inflammatory cytokine expression.Applications Reported: This 85BRD antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This 85BRD antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation and Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols section (Protocol A: Two step protocol for (cytoplasmic) intracellular proteins). This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-13
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P35225
Interleukin-13, ALRH antibody, NC30 antibody, P600 antibody
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
85BRD
PE
IL13
Liquid
IL13
IgG2b, kappa, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3596
Videos
Documents

For Research Use Only.