Learn More
IL-12 p35 Monoclonal Antibody (4D10p35), eFluor™ 660, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£108.00 - £263.00
Specifications
Antigen | IL-12 p35 |
---|---|
Clone | 4D10p35 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15520037
|
Affymetrix eBioscience
50-7352-80 |
25 μg |
£108.00
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15530037
|
Affymetrix eBioscience
50-7352-82 |
100 μg |
£263.00
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This 4D10p35 monoclonal antibody reacts with mouse p35, a component of two of the four members of the interleukin (IL)-12 family of cytokines. The IL-12 family members are heterodimeric cytokines composed of an alpha-chain, such as p35, and a beta-chain, such as p40. The alpha-chain proteins are thought to be the limiting factors for secretion of IL-12 family member cytokines, as expression of the alpha-chain is more restricted than that of the beta-chain proteins, and the two subunits may need to be co-expressed in order for cytokine secretion to occur. IL-12 family members are secreted primarily by antigen-presenting cells in response to various TLR agonists and other inflammatory stimuli. The exact types of stimuli present and kinetics of stimulation determine the balance of IL-12 family member production. The p35 protein was first identified as part of the IL-12 cytokine, which is also known as IL-12p70. Secretion is induced by IFN gamma and agonists of TLR3, 4, or 7 and is inhibited by Th2 cytokines. IL-12 is essential for the differentiation and maintece of Th1 cells. The other subunit, p40, is expressed in much higher quantities than p35, and may be secreted independently as a monomer or homodimer. Free p40 may act as an antagonist of IL-12 function. More recently, p35 was also found to dimerize with EBI3 (Epstein-Barr Virus-Induced Gene 3) to form IL-35.
IL-12 p35 Monoclonal antibody specifically detects IL-12 p35 in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-12 p35 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35 | |
IL12A | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
4D10p35 | |
Flow Cytometry | |
eFluor 660 | |
Rat | |
Mouse | |
P43431 | |
16159 | |
Mouse IL-35-Fc recombinant protein | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL12A |
For Research Use Only.