missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-12/IL-23 p40 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | IL-12/IL-23 p40 |
---|---|
Clone | 116224 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Porcine samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
IL-12/IL-23 p40 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Chinese hamster ovary cell line CHO-derived recombinant porcine IL-12 p70 heterodimer | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects the p40 subunit of porcine IL-12/IL-23 in ELISAs and Western blots. In ELISAs, this antibody shows 25-35% cross-reactivity with recombinant human IL-12 and IL-23 and no cross-reactivity with recombinant mouse IL-12 or IL-23. |
116224 | |
ELISA | |
Unconjugated | |
Mouse | |
Porcine | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only