missing translation for 'onlineSavingsMsg'
Learn More

IL-12/IL-23 p40, PE, clone: C8.6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7129-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229299

  • £94.27 / 50µg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The C8.6 antibody reacts with the p40 subunit of human IL-12 p70 and human IL-23. IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits.Applications Reported: The C8.6 antibody has been reported for use in intracellular flow cytometric analysis.Applications Tested: This C8.6 antibody is offered in 2 formats: - μg size: has been tested by intracellular flow cytometric analysis. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated and tested by intracellular flow cytometric analysis. This can be used at test size: 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. The pre-titrated test size contains BSA and/or gelatin for protein stabilization.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-12/IL-23
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P29460
IL12B, Il12p40, interleukin 12B, RP23-388G23.1, Il-12b, Il-12p40, p40, IL-12 p40, IL-23 subunit p40, IL12P40
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
C8.6
PE
IL12B
Conjugated
IL12B
IgG1, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3593
Videos

For Research Use Only.