missing translation for 'onlineSavingsMsg'
Learn More

IL-12/IL-23 p40, eFluor 660, clone: eBioHP40 (HP40, HP-40), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-7235-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15560027

  • £81.72 / 25 tests


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Description

Description

Description: The eBioHP40 antibody reacts with the p40 subunit of human IL-12 p70, and human IL-23. IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and is secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits.The eBioHP40 antibody has slightly lower background staining than C8.6 and recognizes a different epitope.Applications Reported: This eBioHP40 (HP40, HP-40) antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This eBioHP40 (HP40, HP-40) antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of cultured human dendritic cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-12/IL-23
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P29460
IL12B, Il12p40, interleukin 12B, RP23-388G23.1, Il-12b, Il-12p40, p40, IL-12 p40, IL-23 subunit p40, IL12P40
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioHP40 (HP40, HP-40)
eFluor 660
IL12B
Conjugated
IL12B
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3593
Videos

For Research Use Only.