IL-12/IL-23 Mouse anti-Human, eFluor 660, Clone: C8.6, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 50-7129-42

100TEST Anti-Human IL-12/IL-23 p40 eFluor 660

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only.

Product Code. 15550017

Quantity Price
1 £ 278.6 / 100 tests
Estimated Shipment date
from Supplier 21-02-2017
Add to basket


Description and Specification

Specification

Antigen IL-12/IL-23
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias Interleukin-12/Interleukin-23, p40/70
Host Species Mouse
Applications Flow Cytometry (Intracellular Staining)
Isotype IgG1
Quantity 100 tests
Regulatory Status RUO
Species Reactivity Human
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone C8.6
Concentration 5μL (0.03μg)/test
Conjugate eFluor 660

The C8.6 antibody reacts with the p40 subunit of human IL-12 p70 and human IL-23. IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits.