missing translation for 'onlineSavingsMsg'
Learn More

IL-12/IL-23 p40, eFluor 450, clone: C8.6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-7129-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15559806

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The C8.6 antibody reacts with the p40 subunit of human IL-12 p70 and human IL-23. IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits.
Specifications

Specifications

IL-12/IL-23
C8.6
eFluor 450
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-12/Interleukin-23, p40/70
IgG1, kappa
RUO
Primary
Human
Videos

For Research Use Only.