missing translation for 'onlineSavingsMsg'
Learn More

IL-10 Rat anti-Human, PerCP-eFluor™ 710, Clone: JES3-9D7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-7108-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15599346

  • £213.00 / 100 tests

EU Stock 14

Please to purchase this item. Need a web account? Register with us today!


Description

Description

00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. Interleukin 10 (IL-10, CSIF) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 is an 18.6 kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein (BCRFI). The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including IFN-gamma, TNF-alpha, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells, enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Binds to a retinoid X receptor (RXR) responsive element from the cellular retinol-binding protein II promoter (CRBPII-RXRE). IL-10 inhibits the 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element. Knockout studies in mice suggest the function of this cytokine as an essential immunoregulator in the intestinal tract.
Specifications

Specifications

IL-10
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P22301
Interleukin-10, CSIF, IL-10, IL10A, TGIF
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
JES3-9D7
PerCP-eFluor™ 710
IL10
Liquid
IL10
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3586
Videos
Documents

For Research Use Only.