missing translation for 'onlineSavingsMsg'
Learn More

IL-10, Alexa Fluor 488, clone: JES3-9D7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-7108-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15510247

  • £159.14 / 100 tests


EU Stock 7
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The JES3-9D7 monoclonal antibody reacts with human interleukin-10 (IL-10).Applications Reported: This JES3-9D7 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This JES3-9D7 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-10
JES3-9D7
Alexa Fluor 488
Conjugated
Rat
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-10
IgG1
RUO
Primary
Human
Videos