missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine IL-10 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | IL-10 |
---|---|
Clone | 138111 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
IL-10 Monoclonal specifically detects IL-10 in Canine samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
IL-10 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
IL10 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
138111 | |
ELISA | |
Unconjugated | |
Mouse | |
Canine | |
XP_855560 | |
3586 | |
E. coli-derived recombinant canine IL-10 Ser20-Ile179 Accession # XP_855560 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Delects canine IL-10 in ELISAs. In ELISAs, this antibody does not cross-react with recombinant feline IL-10, recombinant human (rh) IL-10, recombinant mouse (rm) IL-10, recombinant porcine IL-10, rhIL-10 sR, rmIL-10 sR, or recombinant rat IL-10. |
For Research Use Only