missing translation for 'onlineSavingsMsg'
Learn More

Human IL-1 RAcP/IL-1 R3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications

POA

Specifications

Antigen IL-1 RAcP/IL-1 R3
Concentration LYOPH
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
View More Specs

Description

Description

IL-1 RAcP/IL-1 R3 Polyclonal specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications

Specifications

IL-1 RAcP/IL-1 R3
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Polyclonal
Goat
Human
C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 R3, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, IL1RAcP, IL-1RAcP, IL1RAP, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta
IL1RAP
IgG
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
LYOPH
Western Blot, Flow Cytometry, CyTOF
Unconjugated
RUO
Q9NPH3
3556
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-1 RAcP/IL-1 R3 Ser21-Glu359 Accession # Q9NPH3
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects human IL-1 RAcP/IL-1 R3 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse IL-1 RAcP is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only