missing translation for 'onlineSavingsMsg'
Learn More

IL-1 alpha, FITC, clone: 364/3B3-14, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-7118-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15228359

  • £74.42 / 50µg


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 364/3B3-14 antibody reacts with human interleukin-1alpha.Applications Reported: The 364/3B3-14 antibody has been reported for use as capture antibody in a human IL-1 alpha ELISA and for intracellular staining for flow cytometric analysis.Applications Tested: This 364/3B3-14 antibody is offered in 2 formats: - μg size: has been tested by intracellular staining and flow cytometric analysis. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated and tested by intracellular staining and flow cytometric analysis. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-1 alpha
364/3B3-14
FITC
Conjugated
Murine
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
0.5mg/mL; 5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-1 alpha
IgG1
RUO
Primary
Human
Videos