missing translation for 'onlineSavingsMsg'
Learn More

Mouse IGSF4A/SynCAM1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications

POA

Specifications

Antigen IGSF4A/SynCAM1/CADM1
Concentration LYOPH
Dilution Western Blot 0.1 ug/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Description

Description

IGSF4A/SynCAM1/CADM1 Polyclonal specifically detects IGSF4A/SynCAM1/CADM1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

IGSF4A/SynCAM1/CADM1
Western Blot 0.1 ug/mL
Polyclonal
Goat
Mouse
BL2, Cadm1, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, SynCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1
Ikbkg
IgG
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
LYOPH
Western Blot
Unconjugated
RUO
NP_061240
23705
Mouse myeloma cell line NS0-derived recombinant mouse IGSF4A/SynCAM1 Ile42-Asp376 Accession # NP_061240
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects mouse IGSF4A/SynCAM1 in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only