missing translation for 'onlineSavingsMsg'
Learn More

Rat IgM, PE-eFluor 610, clone: eBRM, Isotype Control, eBioscience™

Rat Isotype Control

£59.00 - £133.91

Specifications

Antigen Rat IgM
Clone eBRM
Target Species Not Applicable
Conjugate PE-eFluor 610
Isotype IgM, kappa, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15570497
View Documents
Affymetrix eBioscience
61-4341-80
25μg £59.00
25µg View alternative products
Save Up to 
»
Estimated Shipment
01-12-2020
Add to basket
 
15580497
View Documents
Affymetrix eBioscience
61-4341-82
100μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Description: The monoclonal rat IgM K is useful as an isotype control immunoglobulin. Applications Reported: Rat IgM Isotype Control PE-eFluor™ 610 has been reported for use in flow cytometric analysis. Applications Tested: Rat IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Rat IgM
Not Applicable
IgM, kappa, kappa
Monoclonal
Control
PBS with 0.09% sodium azide; pH 7.2
RUO
eBRM
PE-eFluor 610
Control, Flow Cytometry
Liquid
Isotype Control
Store at 2-8°C. Do not freeze. Light-sensitive material.

For Research Use Only.