missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Monoclonal Antibody (II/41), PE-eFluor™ 610, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£76.75 - £221.00
Specifications
Antigen | IgM |
---|---|
Clone | II/41 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-eFluor 610 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15540527
|
Affymetrix eBioscience
61-5790-82 |
100 μg |
£221.00
100µg |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15530527
|
Affymetrix eBioscience
61-5790-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
IgM Monoclonal antibody specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.Specifications
IgM | |
Flow Cytometry | |
PE-eFluor 610 | |
Rat | |
Mouse | |
16019 | |
IgG2a κ | |
Affinity chromatography | |
Ighm |
II/41 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Ighm | |
Primary | |
Antibody |
For Research Use Only.