missing translation for 'onlineSavingsMsg'
Learn More

Rat IgM, FITC, clone: eBRM, Isotype Control, eBioscience™

Rat Isotype Control

Brand:  Affymetrix eBioscience 11-4341-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15228219

  • £49.65 / 50µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The monoclonal rat IgM, kappa is useful as an isotype control immunoglobulin.Applications Reported: Rat IgM Isotype Control FITC has been reported for use in flow cytometric analysis.Applications Tested: Rat IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgM
Control
FITC
Conjugated
Rat
Affinity chromatography
RUO
Isotype Control
Flow Cytometry
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgM, IgM Mu
IgM, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos

For Research Use Only.