Learn More
IgM Monoclonal Antibody (II/41), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 48-5790-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The II/41 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated II/41 can be used as a detection secondary for mouse IgM. Applications Reported: This II/41 antibody has been reported for use in flow cytometric analysis. Applications Tested: This II/41 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
IgM Monoclonal antibody specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.Specifications
IgM | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P01872 | |
Ighm | |
Affinity chromatography | |
RUO | |
16019 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
II/41 | |
eFluor 450 | |
Ighm | |
AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.