Learn More
IgM Monoclonal Antibody (SA-DA4), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£98.75 - £250.00
Specifications
Antigen | IgM |
---|---|
Clone | SA-DA4 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15519866
|
Affymetrix eBioscience
48-9998-42 |
100 Tests |
£250.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15509866
|
Affymetrix eBioscience
48-9998-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The SA-DA4 monoclonal antibody reacts with the mu heavy chain of human IgM. It does not react with other classes of human immunoglobulin including IgD, IgG, or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of mature B cells. Applications Reported: This SA-DA4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SA-DA4 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
IgM Monoclonal antibody specifically detects IgM in Human samples. It is validated for Flow Cytometry.Specifications
IgM | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P01871 | |
IGHM | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IGHM |
SA-DA4 | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human | |
3507 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.