missing translation for 'onlineSavingsMsg'
Learn More

IgM Mouse anti-Human, APC, Clone: SA-DA4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-9998-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15578296

  • £187.00 / 100 tests

EU Stock 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications

Specifications

IgM
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01871
IGHM
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3507
Flow Cytometry
SA-DA4
APC
IGHM
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos
Documents

For Research Use Only.