Learn More
Epredia™ IgM Antibody
Designed for immunofluorescence staining, Epredia™ IgM, FITC Labeled Antibody allows rapid and inexpensive assessment of tissues.
Brand: Epredia™ RB-1922-R2
Code : LD
Additional Details : Weight : 0.00400kg
Description
Host Species: Rabbit
Species Reactivity: Reacts with human μ-chain IgM.
Immunogen: IgM from human serum
Positive Control: Normal tonsil fixed and processed in the same manner as the test specimen
Cellular Localization: Aggregates found in tonsil, lymph node, Peyer's patch and spleen
Recommended for:
- Immunofluorescence
Specifications
IgM | |
Polyclonal | |
FITC | |
IgM from human serum | |
IVD | |
Primary | |
Prediluted |
Immunofluorescence | |
Polyclonal | |
Rabbit | |
2 mL | |
Cancer and Tumor Biology | |
Human |