Learn More
IgM[a] Mouse anti-Mouse, Brilliant Violet 786, Clone: DS-1, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 743890
Additional Details : Weight : 0.01000kg
Description
The DS-1 antibody reacts specifically with mouse IgM of Igh-C[a] /and related haplotypes (e.g., BALB/c, C58, CBA, C3H/Bi, C3H/He, DBA/1, DBA/2). It does not react with IgM of Igh-C[b], Igh-C[e] /, or related hapltypes (e.g., C57BL, SJL, A, AKR, NZB). DS-1 antibody does not react with free μ heavy chain in vitro or in the cytoplasm of pre-B lymphocytes, which lack Ig light chain. It has not been shown to stimulate B-cell proliferation.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.Specifications
IgM[a] | |
DS-1 | |
Brilliant Violet 786 | |
Igh-6a | |
Mouse NMRI Monoclonal Antibody TC31 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |