missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IGHG4 Recombinant Human Protein (108 Ser/Pro)

Expressed from DNA sequence encoding human IgG4 Fc region (Glu 99- Lys 327, 108Ser/Pro)

Brand:  Sino Biological™ 13505HNAH50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15286335

  • £90.31 / 50µg


Estimated Shipment
15-03-2018
Add to basket

Description

Description

  • Formulation: Lyopihilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH: 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
3503
≥90% by SDS-PAGE
Human
<1 EU/μg
IGHG4
Other Proteins
IGHG4 Recombinant Human Protein
Lyophilized
25.5kd
50μg
Truncated, Recombinant
IGHG4 Recombinant Human Protein (108 Ser/Pro)
Assay Standard
Mammalian
Untagged
Videos

For Research Use Only. Not for use in diagnostic procedures.