missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgG3 Isotype Control, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen™ MG300
58.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.50000kg
Description
Mouse IgG3 Isotype Control for Flow
Mouse IgG3 Control antibody specifically detects Mouse IgG3 in Not Applicable samples. It is validated for Flow Cytometry.Specifications
Mouse IgG3 | |
Unconjugated | |
IgG3; Ighg3; Immunoglobulin G; Immunoglobulin G3; Immunoglobulin heavy constant gamma 3; ImmunoglobulinG; ImmunoglobulinG3 | |
purified | |
ASR | |
Isotype Control (Primary) | |
2-8°C | |
IgG3 | |
Wet Ice |
Flow Cytometry | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
Not Applicable | |
Liquid | |
50 Tests |