missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG3, clone: B10, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 14-4742-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15237377

  • £30.74 / 50µg


Estimated Shipment
01-05-2018
Add to basket

Description

Description

Description: The mouse IgG3 lambda is useful as an isotype control immunoglobulin.Applications Reported: This mouse IgG3 isotype control has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis.Applications Tested: This mouse IgG3 isotype control has been tested by flow cytometric analysis of normal human peripheral blood cells. Use the isotype control at the same concentration as the experimental antibody.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG3
Control
Unconjugated
Purified
Murine
Affinity chromatography
RUO
Isotype Control
Immunohistochemistry
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
IgG3
IgG3, lambda
50μg
Store at 2-8°C.
Monoclonal
Videos

For Research Use Only.