missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG3, κ, Mouse, BV786, Clone: J606, Isotype Control, BD Horizon™
Mouse Monoclonal Isotype Control
Brand: BD Biosciences 565361
Additional Details : Weight : 0.01000kg
Description
IgG3, κ Monoclonal antibody specifically detects IgG3, κ in samples. It is validated for Isotype control, Flow cytometry.Specifications
IgG3, κ | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Primary | |
IgG3 κ |
Flow Cytometry | |
J606 | |
Brilliant Violet 786 | |
Anti-Fructosan | |
50 μg | |
Cell Biology | |
Purified |
For Reseach Use Only.