Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 506, eBioscience™, Invitrogen™
Rat Isotype Control
£66.25 - £151.00
Specifications
Antigen | Rat IgG2b kappa |
---|---|
Clone | eB149/10H5 |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | eFluor 506 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15530677
|
Affymetrix eBioscience
69-4031-80 |
25 μg |
£66.25
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15540677
|
Affymetrix eBioscience
69-4031-82 |
100 μg |
£151.00
100µg |
Estimated Shipment: 30-04-2024
Log in to see stock. |
|||||
Description
Description: The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eB149/10H5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eB149/10H5 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody. eFluor® 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b kappa Control antibody specifically detects Rat IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG2b kappa | |
0.2 mg/mL | |
eFluor 506 | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eB149/10H5 | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.