missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2b, κ, Mouse, FITC, Clone: 27-35, Isotype Control, BD Pharmingen™
Mouse Monoclonal Isotype Control
Brand: BD Biosciences 565379
Additional Details : Weight : 0.01000kg
Description
IgG2b, κ Monoclonal antibody specifically detects IgG2b, κ in samples. It is validated for Flow cytometry (Intracellular Staining)), Isotype control.Specifications
IgG2b, κ | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Primary | |
IgG2b κ |
Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
27-35 | |
FITC | |
Anti-Dansyl | |
50 μg | |
Cell Biology | |
Purified |
For Reseach Use Only.