missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat IgG2b Secondary Antibody, eFluor™ 570, eBioscience™, Invitrogen™

Mouse Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 41-4815-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15528946

  • £94.00 / 25µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody R2B-7C3 recognizes Rat IgG2b antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2b or any mouse isotype antibodies. Applications Reported: This R2B-7C3 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This R2B-7C3 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded sections that have been stained first with a rat IgG2b anti-human antibody. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm

Rat IgG2b Monoclonal antibody specifically detects Rat IgG2b in Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).
Specifications

Specifications

Rat IgG2b
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Affinity chromatography
RUO
Rat
Liquid
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
R2B-7C3
eFluor 570
Mouse
25 μg
Secondary
4° C, store in dark, DO NOT FREEZE!
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.