Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Isotype Control
£77.00 - £171.00
Specifications
Antigen | Rat IgG2b kappa |
---|---|
Clone | eB149/10H5 |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | PerCP-eFluor 710 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15371310
|
Affymetrix eBioscience
46-4031-80 |
25 μg |
£77.00
25µg |
Estimated Shipment: 04-04-2024
Log in to see stock. |
|||||
15381310
|
Affymetrix eBioscience
46-4031-82 |
100 μg |
£171.00
100µg |
In Stock
Log in to see stock. |
|||||
Description
Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: PerCP-eFluor® 710 Rat IgG2b Isotype Control has been reported for use in flow cytometric analysis. Applications Tested: PerCP-eFluor® 710 Rat IgG2b Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells.Use at the same concentration as experimental antibody. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b kappa Control antibody specifically detects Rat IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG2b kappa | |
0.2 mg/mL | |
PerCP-eFluor 710 | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eB149/10H5 | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.