Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 25-4031-81
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: PE-Cy7 Rat IgG2b Isotype Control has been reported for use in flow cytometric analysis. Applications Tested: Rat IgG2b K Isotype Control PE-Cyanine7 has been tested by flow cytometric analysis of mouse splenocytes. It should be used at the same concentration as the experimental antibody. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b kappa Control antibody specifically detects Rat IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG2b kappa | |
eB149/10H5 | |
PE-Cyanine7 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
50 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2b κ |
For Research Use Only.