Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 660, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 50-4031-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: eFluor® 660 Rat IgG2b Isotype Control has been reported for use in flow cytometric analysis and immunohistochemical staining. Applications Tested: eFluor® 660 Rat IgG2b Isotype Control has been tested by flow cytometric analysis of mouse splenocytes. This can be used at the same concentration as the experimental antibody. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b kappa Control antibody specifically detects Rat IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunohistochemistry.Specifications
Rat IgG2b kappa | |
eB149/10H5 | |
eFluor 660 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2b κ |
For Research Use Only.