Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 570, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 41-4031-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This Rat IgG2b K Isotype Control eFluor® 570 has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This Rat IgG2b K Isotype Control eFluor® 570 has been tested by immunocytochemistry on fixed and permeabilized cells. Use isotype control at same concentration as experimental antibody. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Description: The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: This Rat IgG2b K Isotype Control eFluor™ 570 has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry.Applications Tested: This Rat IgG2b K Isotype Control eFluor™ 570 has been tested by immunocytochemistry on fixed and permeabilized cells. Use isotype control at same concentration as experimental antibody.Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable.Excitation: 555 nm; Emission: 570 nmSpecifications
Rat IgG2b kappa | |
eB149/10H5 | |
eFluor 570 | |
IgG2, IgG2b, IgG2bk | |
Affinity chromatography | |
RUO | |
None | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Rat | |
25 μg | |
Isotype Control (Primary) | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
IgG2b κ |
For Research Use Only.