Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 570, eBioscience™, Invitrogen™
Rat Isotype Control
£118.00 - £224.00
Specifications
Antigen | Rat IgG2b kappa |
---|---|
Clone | eB149/10H5 |
Conjugate | eFluor 570 |
Form | Liquid |
Host Species | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15508936
|
Affymetrix eBioscience
41-4031-82 |
100 μg |
£224.00
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15598926
|
Affymetrix eBioscience
41-4031-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This Rat IgG2b K Isotype Control eFluor® 570 has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This Rat IgG2b K Isotype Control eFluor® 570 has been tested by immunocytochemistry on fixed and permeabilized cells. Use isotype control at same concentration as experimental antibody. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Rat IgG2b kappa Control antibody specifically detects Rat IgG2b kappa in Not Applicable samples. It is validated for Control, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Rat IgG2b kappa | |
eFluor 570 | |
Rat | |
IgG2b κ | |
Affinity chromatography |
eB149/10H5 | |
Liquid | |
RUO | |
Isotype Control (Primary) |
For Research Use Only.