missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2b kappa, APC-eFluor 780, clone: eB149/10H5, Isotype Control, eBioscience™

Rat Isotype Control

Brand:  Affymetrix eBioscience 47-4031-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15311650

  • £49.84 / 25µg


EU Stock 4
Add to basket

Description

Description

Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: APC-eFluor® 780 Rat IgG2b Isotype Control (APC-Alexa Fluor® 750 replacement) has been reported for use in flow cytometric analysis.Applications Tested: Rat IgG2b K Isotype Control APC-eFluor® 780 has been tested by flow cytometric analysis of mouse splenocytes. Use isotype control at the same concentration as experimental antibody.APC-eFluor® emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG2b, K
Control
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
IgG2, IgG2b, IgG2bk
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Monoclonal
Flow Cytometry
eB149/10H5
APC-eFluor 780
Conjugated
Rat
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.