missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2b kappa, PerCP-eFluor™ 710, clone: eBMG2b, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 46-4732-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15381320

  • £64.55 / 25µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin.Applications Reported: Mouse IgG2b Isotype Control PerCP-eFluor® 710 has been reported for use in flow cytometric analysis.Applications Tested: Mouse IgG2b K Isotype Control PerCP-eFluor® 710 has been tested by flow ctometric analysis of normal human peripheral blood cells. Use isotype control at the same concentration as experimental antibody.PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG2b, K
Control
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG2, IgG2b, IgG2bk
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
eBMG2b
PerCP-eFluor 710
Conjugated
Murine
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.